Merck
  • Synthesis of β-Glycosyl Amides from N-Glycosyl Dinitrobenzenesulfonamides.
Synthesis of β-Glycosyl Amides from N-Glycosyl Dinitrobenzenesulfonamides.
Journal of carbohydrate chemistry (2013-01-26)
Vishwanath Gaitonde, Steven J Sucheck
ABSTRACT
The N-glycosyl-2,4-dinitrobenzenesulfonamides were accessed via benzoyl-protected β-glycosyl azides. The azides were reduced with Adams' catalyst to the corresponding amines. The glycosylamines were sulfonated with 2,4-dinitrobenzenesulfonyl chloride to form N-glycosyl-2,4-dinitrobenzenesulfonamides in moderate yields. β-Glycosyl amides were then prepared in 67 - 81 % yields by treatment of the sulfonamides with thioacetic acid and cesium carbonate. The conversion of the glycosylsulfonamide to the glycosyl amide proceeded with high stereoselectivity.