Merck
  • Serum- and glucocorticoid-regulated kinase 1 is required for nuclear export of the ribonucleoprotein of influenza A virus.
Serum- and glucocorticoid-regulated kinase 1 is required for nuclear export of the ribonucleoprotein of influenza A virus.
Journal of virology (2013-03-15)
Judith G Alamares-Sapuay, Luis Martinez-Gil, Silke Stertz, Matthew S Miller, Megan L Shaw, Peter Palese
ABSTRACT
We previously performed a small interfering RNA (siRNA) screen and identified serum- and glucocorticoid-regulated kinase 1 (SGK1) as a host factor required for influenza A virus replication. However, the role of SGK1 in the influenza viral life cycle has never been examined. In this study, we demonstrate that SGK1 is required for optimal replication of influenza virus, using the SGK1 inhibitor GSK 650394 and SGK1-specific siRNAs. We also demonstrate that SGK1 is required for viral ribonucleoprotein nuclear export.
Sigma-Aldrich
GSK-650394, ≥98% (HPLC)