Merck
  • GFI1B, EVI5, MYB--additional genes that cooperate with the human BCL6 gene to promote the development of lymphomas.
GFI1B, EVI5, MYB--additional genes that cooperate with the human BCL6 gene to promote the development of lymphomas.
Blood cells, molecules & diseases (2013-08-06)
Beverly W Baron, John Anastasi, Juraj Bies, Poluru L Reddy, Loren Joseph, Michael J Thirman, Kristen Wroblewski, Linda Wolff, Joseph M Baron
ABSTRACT
The BCL6 gene, which is expressed in certain B- and T-cell human lymphomas, is involved with chromosomal rearrangements and mutations in a number of these neoplasms. Lymphomagenesis is believed to evolve through a multi-step accumulation of genetic alterations in these tumors. We used retroviral insertional mutagenesis in transgenic mice expressing the human BCL6 transgene in order to identify genes that cooperate with BCL6 during lymphomatous transformation. We previously reported PIM1 as the most frequently recurring cooperating gene in this model. We now report three newly identified cooperating genes-GFI1B, EVI5, and MYB-that we identified in the lymphomas of retroviral-injected BCL6 transgenic mice (but not in retroviral-injected non-transgenic controls); mRNA and protein expression of GFI1B and EVI5 were decreased in the murine tumors, whereas MYB mRNA and protein expression were increased or decreased. These findings correlated with protein expression in human lymphomas, both B- and T-cell. Improved therapy of lymphomas may necessitate the development of combinations of drugs that target the alterations specific to each neoplasm.
Sigma-Aldrich
Anti-EVI5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution