- Wnt antagonist SFRP1 functions as a secreted mediator of senescence.
Wnt antagonist SFRP1 functions as a secreted mediator of senescence.
Molecular and cellular biology (2012-08-29)
David J Elzi, Meihua Song, Kevin Hakala, Susan T Weintraub, Yuzuru Shiio
PMID22927647
ABSTRACT
Cellular senescence has emerged as a critical tumor suppressive mechanism in recent years, but relatively little is known about how senescence occurs. Here, we report that secreted Frizzled-related protein 1 (SFRP1), a secreted antagonist of Wnt signaling, is oversecreted upon cellular senescence caused by DNA damage or oxidative stress. SFRP1 is necessary for stress-induced senescence caused by these factors and is sufficient for the induction of senescence phenotypes. We present evidence suggesting that SFRP1 functions as a secreted mediator of senescence through inhibition of Wnt signaling and activation of the retinoblastoma (Rb) pathway and that cancer-associated SFRP1 mutants are defective for senescence induction.
Sigma-Aldrich
sFRP-1 human, recombinant, expressed in HeLa cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture