- Long-term antibiotic therapy for balanitis xerotica obliterans.
Long-term antibiotic therapy for balanitis xerotica obliterans.
Journal of the American Academy of Dermatology (1999-01-28)
W B Shelley, E D Shelley, M A Grunenwald, T J Anders, A Ramnath
PMID9922014
ABSTRACT
In an open uncontrolled study of 3 patients with balanitis xerotica obliterans we have observed significant improvement after long-term systemic antibiotic therapy. Two of the patients noticed softening of the skin as well as disappearance of pruritus, tenderness, and inflammatory changes within 3 weeks of receiving oral and intramuscular penicillin. The third patient experienced the same degree of improvement after a regimen of dirithromycin, 500 mg/day. Stopping the antiobiotic for 1 month resulted in relapse, with improvement again on resumption.