Merck
  • Butyric acid and pivaloyloxymethyl butyrate, AN-9, a novel butyric acid derivative, induce apoptosis in HL-60 cells.
Butyric acid and pivaloyloxymethyl butyrate, AN-9, a novel butyric acid derivative, induce apoptosis in HL-60 cells.
Journal of cancer research and clinical oncology (1997-01-01)
Y Zimra, L Wasserman, L Maron, M Shaklai, A Nudelman, A Rephaeli
ABSTRACT
A novel butyric acid derivative, pivaloyloxymethyl butyrate, AN-9, was previously shown to be a potent differentiating agent. AN-9 exerts a significant anticancer activity in vitro and in vivo. In all the activities examined, AN-9 was more potent than butyric acid. Here we show that AN-9 and butyric acid induce cell death by apoptosis. Exposure of HL-60 cells to butyric acid and AN-9 decreased cell numbers and induced cell differentiation and the appearance of typical apoptotic features. Induction of apoptosis and/or differentiation by AN-9 and butyric acid was dependent on the concentration and the time of exposure to the drugs. The advantage of AN-9 over butyric acid was further confirmed. Apoptosis induced by AN-9 occurred after a shorter exposure and at lower drug concentrations than that induced by butyric acid. Apoptosis by AN-9 was accompanied by reduction in Bcl-2 expression. Preincubation with antioxidants did not protect HL-60 cells from apoptosis induced by AN-9. HL-60 cells that were induced to differentiate by preincubation with retinoic acid or low AN-9 concentrations were more resistant to apoptosis, induced later by high concentrations of AN-9, than were undifferentiated cells.