Merck
  • The motilin antagonist ANQ-11125 blocks motilide-induced contractions in vitro in the rabbit.
The motilin antagonist ANQ-11125 blocks motilide-induced contractions in vitro in the rabbit.
Biochemical and biophysical research communications (1994-01-28)
T L Peeters, I Depoortere, M J Macielag, R Dharanipragada, M S Marvin, J R Florance, A Galdes
ABSTRACT
Studies on the physiological role of motilin, and more recently, on the relationship between motilin and erythromycin A, have been hampered by the lack of antagonists. We now have discovered such a compound. ANQ-11125 displaces motilin bound to an homogenate of rabbit antral smooth muscle tissue. The dissociation constant (pKd) was 8.16 +/- 0.10. However, ANQ-11125 did not induce contractions of segments of rabbit duodenum, except at high concentrations. In the presence of 1 microM ANQ-11125 the dose response curves of erythromycin-A, De(N-methyl)-N-ethyl-8,9 anhydroerythromycin A 6,9-hemiacetal and motilin were shifted about one log unit to the right, but the responses to ACh and Substance P were unaffected. Schild-analysis showed the competitive nature of the interaction and allowed the calculation of the pA2: 7.03 +/- 0.05 (motilin curves) and 7.55 +/- 0.06 (EM-523 curves). This is the first report of a motilin antagonist. Its properties definitively prove that motilides are motilin agonists.
Product Description
Supelco
Erythromycin A enol ether, analytical standard