Merck
  • Metabolism of unsaturated bile acids and androstanes by human faecal bacteria.
Metabolism of unsaturated bile acids and androstanes by human faecal bacteria.
Journal of steroid biochemistry (1985-06-01)
R W Owen, R F Bilton
ABSTRACT
The metabolism of unsaturated bile acids and androstanes by mixed human faecal cultures has been studied. The reactions observed were mainly reductive. Unsaturated 4-ene-3-oxo and 1,4-diene-3-oxo bile acids were reduced in Ring A. 5 beta-3-Oxo bile acids were reduced to 5 beta-3-hydroxy bile acids. 4-Ene, 1,4-diene and 4,6-diene-3,17-dioxo-androstanes were reduced in Ring A with concomitant reduction of oxo groups to hydroxyl groups. The Gram-negative facultative anaerobic faecal bacteria are implicated in the reductive process, whilst the genus Clostridium does not appear to be important. Inclusion of menadione, a synthetic form of vitamin K, retards the reductive process.