Merck
  • Structure-mutagenicity relationships of chalcones and their oxides in the Salmonella assay.
Structure-mutagenicity relationships of chalcones and their oxides in the Salmonella assay.
Mutation research (1986-03-01)
K A Rashid, C A Mullin, R O Mumma
ABSTRACT
31 p-monosubstituted chalcones (E-1, 3-diphenylpropene-1-one) and the corresponding oxides (E-1-benzoyl-2-phenyloxirane) were tested for mutagenic activity on two strains of Salmonella typhimurium (TA98 and TA100) with and without rat liver microsomal and cytosolic enzymes. Highest mutagenicity (3.0 revertants/nmole in either strain) was seen with the 4-nitrochalcone, especially after S9 activation. Epoxidation, in general, increased the mutagenic activity of the respective chalcone. Benzoyl (4') substituted chalcones and their oxides with an electron-withdrawing substituent (e.g., nitro, fluoro) usually had higher activity than their phenyl (4) substituted counterparts, whereas the converse was the case with electron-donating substituents (e.g., acetamido, methoxy). Further multiple factorial analysis revealed that increasing hydrophilicity as indicated by the Hansch pi parameter, and resonance electronic contributions were more important than other factors including steric terms in explaining the mutagenicity of these compounds. Mutagenic effects of some chalcone oxides, particularly the 4-methoxy derivative, were markedly decreased by S9 treatment. The consequence of the weak-to-moderate mutagenicity of these compounds to dietary intake of hydroxylated and methoxylated chalcones is discussed.