Merck
  • Aspects of pharmaceutical and chemical standardization of patch test materials.
Aspects of pharmaceutical and chemical standardization of patch test materials.
Journal of the American Academy of Dermatology (1989-10-01)
B Kreilgård, J Hansen
ABSTRACT
The choice of vehicle for patch test materials is important for the bioavailability and stability of the allergens. Examples are given from the standardization of the TRUE Test and include caine mix, Kathon CG, and fragrance mix.
Sigma-Aldrich
7-Hydroxycitronellal, Odorant used in allergy studies
Sigma-Aldrich
Hydroxycitronellal, ≥95%, FCC, FG