Merck
  • Prediction of late disease recurrence and extended adjuvant letrozole benefit by the HOXB13/IL17BR biomarker.
Prediction of late disease recurrence and extended adjuvant letrozole benefit by the HOXB13/IL17BR biomarker.
Journal of the National Cancer Institute (2013-07-03)
Dennis C Sgroi, Erin Carney, Elizabeth Zarrella, Lauren Steffel, Shemeica N Binns, Dianne M Finkelstein, Jackie Szymonifka, Atul K Bhan, Lois E Shepherd, Yi Zhang, Catherine A Schnabel, Mark G Erlander, James N Ingle, Peggy Porter, Hyman B Muss, Katherine I Pritchard, Dongsheng Tu, David L Rimm, Paul E Goss
ABSTRACT
Biomarkers to optimize extended adjuvant endocrine therapy for women with estrogen receptor (ER)-positive breast cancer are limited. The HOXB13/IL17BR (H/I) biomarker predicts recurrence risk in ER-positive, lymph node-negative breast cancer patients. H/I was evaluated in MA.17 trial for prognostic performance for late recurrence and treatment benefit from extended adjuvant letrozole. A prospective-retrospective, nested case-control design of 83 recurrences matched to 166 nonrecurrences from letrozole- and placebo-treated patients within MA.17 was conducted. Expression of H/I within primary tumors was determined by reverse-transcription polymerase chain reaction with a prespecified cutpoint. The predictive ability of H/I for ascertaining benefit from letrozole was determined using multivariable conditional logistic regression including standard clinicopathological factors as covariates. All statistical tests were two-sided. High H/I was statistically significantly associated with a decrease in late recurrence in patients receiving extended letrozole therapy (odds ratio [OR] = 0.35; 95% confidence interval [CI] = 0.16 to 0.75; P = .007). In an adjusted model with standard clinicopathological factors, high H/I remained statistically significantly associated with patient benefit from letrozole (OR = 0.33; 95% CI = 0.15 to 0.73; P = .006). Reduction in the absolute risk of recurrence at 5 years was 16.5% for patients with high H/I (P = .007). The interaction between H/I and letrozole treatment was statistically significant (P = .03). In the absence of extended letrozole therapy, high H/I identifies a subgroup of ER-positive patients disease-free after 5 years of tamoxifen who are at risk for late recurrence. When extended endocrine therapy with letrozole is prescribed, high H/I predicts benefit from therapy and a decreased probability of late disease recurrence.
Product Description
Letrozole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Letrozole, ≥98% (HPLC)