Merck
  • Rapid-screening detection of acetildenafils, sildenafils and avanafil by ion mobility spectrometry.
Rapid-screening detection of acetildenafils, sildenafils and avanafil by ion mobility spectrometry.
Journal of pharmaceutical and biomedical analysis (2012-12-25)
Daniel J Mans, Rebecca J Callahan, Jamie D Dunn, Connie M Gryniewicz-Ruzicka
ABSTRACT
Ion mobility spectrometry was used as a rapid screening tool for the detection of acetildenafils, sildenafils and avanafil within adulterated herbal supplement matrices. Acetildenafils show a tendency for partial fragmentation during the desorption/ionization process affording two peaks in the ion mobility spectrum in addition to the intact compound. The fragmentation appears to occur α to the carbonyl group along the CN bond attaching the piperazine moiety, producing a common fragment (K₀=1.0280 cm²V⁻¹s⁻¹) along with the respective piperazine fragment. The sildenafils and avanafil afford one molecular ion peak per compound.
Product Description
Supelco
Vardenafil dihydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Zopiclone-D4 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®