Merck
  • A PKS gene, pks-1, is involved in chaetoglobosin biosynthesis, pigmentation and sporulation in Chaetomium globosum.
A PKS gene, pks-1, is involved in chaetoglobosin biosynthesis, pigmentation and sporulation in Chaetomium globosum.
Science China. Life sciences (2012-12-13)
Yang Hu, Xiaoran Hao, Jing Lou, Ping Zhang, Jiao Pan, Xudong Zhu
ABSTRACT
Chaetomium globosum is one of the most common fungi in nature. It is best known for producing chaetoglobosins; however, the molecular basis of chaetoglobosin biosynthesis is poorly understood in this fungus. In this study, we utilized RNA interference (RNAi) to characterize a polyketide synthase gene, pks-1, in C. globosum that is involved in the production of chaetoglobosin A. When pks-1 was knocked down by RNAi, the production of chaetoglobosin A dramatically decreased. Knock-down mutants also displayed a pigment-deficient phenotype. These results suggest that the two polyketides, melanin and chaetoglobosin, are likely to share common biosynthetic steps. Most importantly, we found that pks-1 also plays a critical role in sporulation. The silenced mutants of pks-1 lost the ability to produce spores. We propose that polyketides may modulate cellular development via an unidentified action. We also suggest that C. globosum pks-1 is unique because of its triple role in melanin formation, chaetoglobosin biosynthesis and sporulation. This work may shed light on chaetoglobosin biosynthesis and indicates a relationship between secondary metabolism and fungal morphogenesis.
Sigma-Aldrich
Chaetoglobosin A, from Chaetomium globosum, ≥98% (HPLC)