Merck
  • A temperature sensor based on CdTe quantum dots-layered double hydroxide ultrathin films via layer-by-layer assembly.
A temperature sensor based on CdTe quantum dots-layered double hydroxide ultrathin films via layer-by-layer assembly.
Chemical communications (Cambridge, England) (2012-12-13)
Ruizheng Liang, Rui Tian, Wenying Shi, Zhihui Liu, Dongpeng Yan, Min Wei, David G Evans, Xue Duan
ABSTRACT
Ordered ultrathin films based on alternate assembly of CdTe QDs and layered double hydroxide monolayers have been fabricated, which can be used as a photoluminescence temperature sensor with dual-parameter signals and high response sensitivity.