- [Effect of afobazole on the psychophysiological state of healthy volunteers].
[Effect of afobazole on the psychophysiological state of healthy volunteers].
Eksperimental'naia i klinicheskaia farmakologiia (2012-01-12)
N G Bogdan, N V Kolotilinskaia, S A Nadorov, M A Iarkova, B A Badyshtov
PMID22232907
ABSTRACT
The influence of the selective anxiolytic drug afobazole in a single dose of 10 mg on the psychophysiological functions of healthy volunteers was studied in laboratory experiments. It is established that afobazole optimizes some psychophysiological characteristics of stress-labile individuals and has no negative influenceon the parameters of attention, psychomotor reaction a nd speed of decision making.
Product Description
Supelco
Phenazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®