Merck
  • Effects of sulfhydryl-modifying reagents, 3-nitro-2-pyridinesulfenyl compounds, on the coupling between inhibitory receptors and GTP-binding proteins Gi/Go in rat brain membranes.
Effects of sulfhydryl-modifying reagents, 3-nitro-2-pyridinesulfenyl compounds, on the coupling between inhibitory receptors and GTP-binding proteins Gi/Go in rat brain membranes.
Molecular pharmacology (1990-08-01)
Y Kitamura, S Imai, R Matsueda, Y Nomura
ABSTRACT
To gain insight into the coupling mechanism of inhibitory receptors, 5-hydroxytryptamine1A receptors and alpha 2-adrenoceptors, with GTP-binding proteins (G proteins) in the central nervous system, we examined the effects of two 3-nitro-2-pyridinesulfenyl compounds, S-(3-nitro-2-pyridinesulfenyl)-L-cysteine [Cys(Npys)] and N-t-butoxy-carbonyl-S-(3-nitro-2-pyridinesulfenyl)-L-cysteine [Boc-Cys(Npys)], on 1) specific binding of [3H]8-hydroxy-2-(di-n-propylamino)tetralin (8-OH-DPAT) (5-hydroxytryptamine1A agonist) and [3H]clonidine (alpha 2-agonist) to rat brain membranes, 2) [35S]guanosine 5'-O-(3-thio)triphosphate (GTP gamma S) binding, and 3) pertussis toxin (islet-activating protein) (IAP)-catalyzed ADP-ribosylation of purified Go (an IAP-sensitive G protein present in abundance in the mammalian brain). Treatment with Cys(Npys) led to decreased [3H]8-OH-DPAT and [3H]clonidine binding, similar to the inhibitory effects of IAP and N-ethylmaleimide (NEM) on such binding. However, further treatment of Cys(Npys)-pretreated membranes with dithiothreitol completely abolished the inhibitory effect of Cys(Npys) on the binding of both ligands. On the other hand, treatment with Boc-Cys(Npys) inhibited the effect of several GTP analogs (GTP gamma S, guanylyl-imidodiphosphate, guanylyl)-(beta, gamma-methylene)-diphosphate, and GTP) on [3H]8-OH-DPAT and [3H]clonidine binding. Dithiothreitol and mercaptoethanol treatment of Boc-Cys(Npys)-pretreated membranes did not lead to a recovery of the effect of GTP analogs on agonist binding. Regardless of the presence or absence of GTP gamma S, agonist binding to Boc-Cys(Npys)-pretreated membranes was decreased by further addition of NEM or Cys(Npys). Cys(Npys) blocked [35S]GTP gamma S binding as well as IAP-catalyzed ADP-ribosylation in purified Go. In contrast, Boc-Cys(Npys) partially inhibited ADP-ribosylation and did not affect [35S]GTP gamma S binding. These results suggested that Cys(Npys) modifies the receptor-coupling domain in G proteins, followed by the uncoupling of inhibitory receptors from G proteins, similar to the effects of NEM and IAP. Boc-Cys(Npys), however, seems to stabilize the coupling state between the receptors and G proteins, thus abolishing the GTP gamma S effect.