Merck
  • Substituted 2-aminothiazoles are exceptional inhibitors of neuronal degeneration in tau-driven models of Alzheimer's disease.
Substituted 2-aminothiazoles are exceptional inhibitors of neuronal degeneration in tau-driven models of Alzheimer's disease.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2011-06-15)
Irene Lagoja, Christophe Pannecouque, Gerard Griffioen, Stefaan Wera, Veronica Maria Rojasdelaparra, Arthur Van Aerschot
ABSTRACT
A novel series of 2-aminothiazoles with strong protection in an Alzheimer's disease (AD) model comprising tau-induced neuronal toxicity is disclosed. These derivatives can be synthesized in one-pot and a small SAR of the substitution within these series afforded several compounds that counteracted tau-induced cell toxicity at nanomolar concentrations. These congeners therefore have strong potential as possible treatment for Alzheimer's disease and other related tauopathies.
Sigma-Aldrich
2-Aminothiazole, 97%