Merck
  • Enantioselective synthesis of alkaloids from phenylglycinol-derived lactams.
Enantioselective synthesis of alkaloids from phenylglycinol-derived lactams.
Natural product communications (2011-05-13)
Mercedes Amat, Núria Llor, Rosa Griera, Maria Pérez, Joan Bosch
ABSTRACT
This review is focused on recent synthetic achievements and ongoing work in our laboratory using phenylglycinol-derived oxazolopiperidone lactams as starting materials for the enantioselective synthesis of piperidine-containing alkaloids: madangamines, 2,5-disubstituted decahydroquinoline and 1-substituted tetrahydroisoquinoline alkaloids, the indole alkaloids 20S- and 20R-dihydrocleavamine and quebrachamine, and indole alkaloids of the uleine and silicine groups.
Sigma-Aldrich
N-(2-Hydroxyethyl)aniline, 98%