- Near-infrared chromogenic sensing of organotin species by a cyclopalladated azo dye.
Near-infrared chromogenic sensing of organotin species by a cyclopalladated azo dye.
The Analyst (2010-11-11)
Yue-Feng Zhou, Jia-Ni Wang, Shun-Hua Li, Jin-Gou Xu
PMID21063628
ABSTRACT
A simple cyclopalladated complex of 4-(2-thiazolylazo)resorcinol showed a specific red-to-green colour change upon addition of organotin species in the acetonitrile-water medium.