- Occurrence of hydrin 2 (vasotocinyl-Gly), a new hydroosmotic neurohypophyseal peptide, in secretory granules isolated from the frog (Rana esculenta) neurointermediate pituitary.
Occurrence of hydrin 2 (vasotocinyl-Gly), a new hydroosmotic neurohypophyseal peptide, in secretory granules isolated from the frog (Rana esculenta) neurointermediate pituitary.
Neuroendocrinology (1990-02-01)
J Chauvet, Y Rouillé, M T Chauvet, R Acher
PMID2106101
ABSTRACT
Neurohypophyseal secretory granules have been purified from the frog (Rana esculenta) neurointermediate pituitary gland by sucrose gradient centrifugation, and their polypeptide content has been analyzed by reverse-phase high-pressure liquid chromatography. Aside from vasotocin, mesotocin, and their associated neurophysins, hydrin 2 (vasotocinyl-Gly), previously identified in hydrochloric acid extracts, has been recognized. This finding supports the previous suggestion that hydrin 2, a peptide active on the water permeability of frog bladder and frog skin, is a secreted hormone involved in osmoregulation specific to amphibians. Hydrin 2 has not been found in neurosecretory granules of birds such as the goose.