Merck
  • The effect of oxandrolone on voice frequency in growth hormone-treated girls with Turner syndrome.
The effect of oxandrolone on voice frequency in growth hormone-treated girls with Turner syndrome.
Journal of voice : official journal of the Voice Foundation (2010-10-26)
Leonie A Menke, Theo C J Sas, Sophie H L van Koningsbrugge, Maria A J de Ridder, Gladys R J Zandwijken, Bart Boersma, Philippe H Dejonckere, Sabine M P F de Muinck Keizer-Schrama, Barto J Otten, Jan M Wit
ABSTRACT
Oxandrolone (Ox) increases height gain but may also cause voice deepening in growth hormone (GH)-treated girls with Turner syndrome (TS). We assessed the effect of Ox on objective and subjective speaking voice frequency in GH-treated girls with TS. A multicenter, randomized, placebo (Pl)-controlled, double-blind study was conducted. One hundred thirty-three patients were included and treated with GH (1.33 mg/m2/d) from baseline, combined with Pl or Ox in a low (0.03 mg/kg/d) or conventional (0.06 mg/kg/d) dose from the age of 8 years and estrogens from the age of 12 years. Yearly from starting Ox/Pl until 6 months after discontinuing GH+Ox/Pl, voices were recorded and questionnaires were completed. At start, mean (±standard deviation [SD]) voice frequency SD score (SDS) was high for age (1.0±1.2, P<0.001) but normal for height. Compared with GH+Pl, voices tended to lower on GH+Ox 0.03 (P=0.09) and significantly lowered on GH+Ox 0.06 (P=0.007). At the last measurement, voice frequency SDS was still relatively high in GH+Pl group (0.6±0.7, P=0.002) but similar to healthy girls in both GH+Ox groups. Voice frequency became lower than -2 SDS in one patient (3%) on GH+Ox 0.03 and three patients (11%) on GH+Ox 0.06. The percentage of patients reporting subjective voice deepening was similar between the dosage groups. Untreated girls with TS have relatively high-pitched voices. The addition of Ox to GH decreases voice frequency in a dose-dependent way. Although most voice frequencies remain within the normal range, they may occasionally become lower than -2 SDS, especially on GH+Ox 0.06 mg/kg/d.
Sigma-Aldrich
Oxandrolone, ≥98% (HPLC)