Merck
  • The Fos expression in rat brain following electrical stimulation of dura mater surrounding the superior sagittal sinus changed with the pre-treatment of rizatriptan benzoate.
The Fos expression in rat brain following electrical stimulation of dura mater surrounding the superior sagittal sinus changed with the pre-treatment of rizatriptan benzoate.
Brain research (2010-10-12)
Xiaolin Wang, Shengyuan Yu, Zhao Dong, Lei Jiang
ABSTRACT
Fos expression in the brain was systematically investigated by means of immunohistochemical staining after electrical stimulation of the dura mater surrounding the superior sagittal sinus in conscious rats. Fos-like immunoreactive neurons are distributed mainly in the upper cervical spinal cord, spinal trigeminal nucleus caudal part, raphe magnus nucleus, periaqueductal gray, ventromedial hypothalamic nucleus, and mediodorsal thalamus nucleus. With the pre-treatment of intraperitoneal injection of rizatriptan benzoate, the number of Fos-like immunoreactive neurons decreased in the spinal trigeminal nucleus caudal part and raphe magnus nucleus, increased in the periaqueductal gray, and remained unchanged in the ventromedial hypothalamic nucleus and mediodorsal thalamus nucleus. These results provide morphological evidence that the nuclei described above are involved in the development and maintenance of the trigeminovascular headache.
Sigma-Aldrich
Rizatriptan benzoate salt, ≥98% (HPLC)