- Structural characterization and dynamics of globotetraosylceramide in vascular endothelial cells under TNF-alpha stimulation.
Structural characterization and dynamics of globotetraosylceramide in vascular endothelial cells under TNF-alpha stimulation.
In several vascular inflammatory reactions (i.e. immunity and thrombosis) inflammatory mediators lead to the activation of vascular endothelial cells (EC). To date, a number of functional molecules induced on the surface of activated-EC have been identified. We report here that Globotetraosylceramide (Gb4), a glycosphingolipid expressed in EC, is a novel inducible molecule on EC activated by TNF-alpha. The cell surface expression of Gb4 is increased in a time-dependent manner under TNF-alpha stimulation, which shows distinct expression kinetics of major proteins induced by TNF-alpha on EC. MALDI-TOFMS analysis revealed that the enhanced Gb4 predominantly contains C24:0 fatty acid in the ceramide moiety. Isolated caveolae/lipid raft-enriched detergent insoluble membrane domains in activated-EC predominantly contain this molecular species of Gb4. Gb4 containing C16:0 fatty acid in the ceramide moiety, which is known to constitute the major species of Gb4 in plasma, is also found as a major molecular species in EC. These observations indicate that Gb4, especially with very long fatty acid, is enhanced in EC during its inflammatory reaction, and suggest the potential utility of Gb4 as a biomarker for monitoring inflammation status of EC involving its related diseases.