Merck
  • Protonation thermochemistry of selected hydroxy- and methoxycarbonyl molecules.
Protonation thermochemistry of selected hydroxy- and methoxycarbonyl molecules.
The journal of physical chemistry. A (2005-12-22)
Guy Bouchoux, Danielle Leblanc, William Bertrand, Terance B McMahon, Jan E Szulejko, Florence Berruyer-Penaud, Otilia Mó, Manuel Yáñez
ABSTRACT
The gas-phase basicities of a representative set of hydroxy- and methoxycarbonyl compounds (hydroxyacetone, 1, 3-hydroxybutanone, 2, 3-hydroxy-3-methylbutanone, 3, 1-hydroxy-2-butanone, 4, 4-hydroxy-2-butanone, 5, 5-hydroxy-2-pentanone, 6, methoxyacetone, 7, 3-methoxy-2-butanone, 8, 4-methoxy-2-butanone, 9, and 5-methoxy-2-pentanone, 10) were experimentally determined by the equilibrium method using Fourier transform ion cyclotron resonance and high-pressure mass spectrometry techniques. The latter method allows the measurement of proton transfer equilibrium constants at various temperatures and thus the estimate of both the proton affinities and the protonation entropies of the relevant species. Quantum chemical calculations at the G3 and the B3LYP/6-311+G(3df,2p)//6-31G(d) levels of theory were undertaken in order to find the most stable structures of the neutrals 1-10 and their protonated forms. Conformational and vibrational analyses have been done with the aim of obtaining a theoretical estimate of the protonation entropies.
Sigma-Aldrich
Methoxyacetone, 95%