Merck
  • Synthesis of sterically encumbered C10-arylated benzo[h]quinolines using ortho-substituted aryl boronic acids.
Synthesis of sterically encumbered C10-arylated benzo[h]quinolines using ortho-substituted aryl boronic acids.
Organic & biomolecular chemistry (2012-10-17)
Marko Weimar, Matthew J Fuchter
ABSTRACT
The challenging coupling of 10-halobenzo[h]quinolines with ortho-substituted aryl boronic acids has been achieved using Pd(OAc)(2)/P(O)Ph(3) as the catalytic system. High yields were obtained for diversely functionalised substrates under mild reaction conditions.
Sigma-Aldrich
Benzo[h]quinoline, 97%