Merck
  • Direct effects of caffeine and theophylline on p110 delta and other phosphoinositide 3-kinases. Differential effects on lipid kinase and protein kinase activities.
Direct effects of caffeine and theophylline on p110 delta and other phosphoinositide 3-kinases. Differential effects on lipid kinase and protein kinase activities.
The Journal of biological chemistry (2002-07-30)
Lazaros C Foukas, Nathalie Daniele, Chariklia Ktori, Karen E Anderson, Jorgen Jensen, Peter R Shepherd
ABSTRACT
We investigated the effects of methylxanthines on enzymatic activity of phosphoinositide 3-kinases (PI3Ks). We found that caffeine inhibits the in vitro lipid kinase of class I PI3Ks (IC(50) = 75 microm for p110 delta, 400 microm for p110 alpha and p110 beta, and 1 mm for p110 gamma), and theophylline has similar effects (IC(50) = 75 microm for p110 delta, 300 microm for p110 alpha, and 800 microm for p110 beta and p110 gamma) and also inhibits the alpha isoform of class II PI3K (PI3K-C2 alpha) (IC(50) approximately 400 microm). However, four other xanthine derivatives tested (3-isobutyl-1-methylxanthine, 3-propylxanthine, alloxazine, and PD116948 (8-cyclopentyl-1,3-dipropylxanthine)) were an order of magnitude less effective. Surprisingly the triazoloquinazoline CGS15943 (9-chloro-2-(2-furyl)(1,2,d)triazolo(1,5-c)quinazolin-5-amine) also selectively inhibits p110 delta (IC(50) < 10 microm). Caffeine and theophylline also inhibit the intrinsic protein kinase activity of the class IA PI3Ks and DNA-dependent protein kinase, although with a much lower potency than that for the lipid kinase (IC(50) approximately 10 mm for p110 alpha, 3 mm for p110 beta, and 10 mm for DNA-dependent protein kinase). In CHO-IR cells and rat soleus muscle, theophylline and caffeine block the ability of insulin to stimulate protein kinase B with IC(50) values similar to those for inhibition of PI3K activity, whereas insulin stimulation of ERK1 or ERK2 was not inhibited at concentrations up to 10 mm. Theophylline and caffeine also blocked insulin stimulation of glucose transport in CHO-IR cells. These results demonstrate that these methylxanthines are direct inhibitors of PI3K lipid kinase activity but are distinctly less effective against serine kinase activity and thus could be of potential use in dissecting these two distinct kinase activities. Theophylline, caffeine, and CGS15943 may be of particular use in dissecting the specific role of the p110 delta lipid kinase. Finally, we conclude that inhibition of PI3K (p110 delta in particular) is likely explain some of the physiological and pharmacological properties of caffeine and theophylline.
Sigma-Aldrich
CGS-15943, solid