Merck
  • Determination of hair polyamines as N-ethoxycarbonyl-N-pentafluoropropionyl derivatives by gas chromatography-mass spectrometry.
Determination of hair polyamines as N-ethoxycarbonyl-N-pentafluoropropionyl derivatives by gas chromatography-mass spectrometry.
Journal of chromatography. A (2000-12-29)
M H Choi, K R Kim, B C Chung
ABSTRACT
An efficient method is described for the simultaneous determination of hair polyamines, such as 1,3-diaminopropane, putrescine, cadaverine, spermidine and spermine, by gas chromatography-mass spectrometry (GC-MS) with the selected ion-monitoring (SIM) mode. The method is based on the extractive two-phase ethoxycarbonyl (EOC) reaction of amino functions in aqueous solutions combined with subsequent pentafluoropropionyl (PFP) derivatization of the remaining active hydrogen atoms for the direct analysis by GC-SIM-MS. The detection limits for SIM of the polyamines as N-EOC-N-PFP derivatives ranged from 1 to 10 ng/g hair, while their recovery rates varied in the range of 76.42-93.38%. This method demonstrated a good overall accuracy (% bias) and precision (% C.V.) as 3.32-11.05% and 5.88-14.71%, respectively. When applied to 0.6 M HCl extracts of hair samples from 11 healthy men and 19 healthy women, all five polyamines were positively detected at the concentrations of 8.82-871.87 ng/g. Both in median and mean concentrations, the most abundant hair polyamine was spermidine, followed by spermine, putrescine, 1,3-diaminopropane and cadaverine in the male group, while the order of 1,3-diaminopropane and cadaverine was reversed in the female group. The levels of polyamines, except for cadaverine, in hair specimens studied were found to be higher in men than in women.
Sigma-Aldrich
Pentafluoropropionic acid, 97%
Sigma-Aldrich
Sodium pentafluoropropionate, 98%