Merck
  • Single-dose pharmacokinetics of methylphenidate in CYP2D6 extensive and poor metabolizers.
Single-dose pharmacokinetics of methylphenidate in CYP2D6 extensive and poor metabolizers.
Journal of clinical psychopharmacology (2000-06-01)
C L DeVane, J S Markowitz, S W Carson, D W Boulton, H S Gill, Z Nahas, S C Risch
ABSTRACT
Six adults phenotyped as either extensive (N = 4) or poor (N = 2) metabolizers for cytochrome P450 (CYP) 2D6 were given a 10-mg oral dose of methylphenidate (MPH) on two separate occasions with and without quinidine, a potent CYP2D6 inhibitor. Quinidine had no significant effect on the pharmacokinetics of either MPH or ritalinic acid, its major metabolite, in either group of CYP2D6 metabolizers. These data suggest a lack of involvement of CYP2D6 in the metabolism of MPH. Drugs that are inhibitors of CYP2D6 when taken concurrently with MPH should not affect its plasma concentration.
Sigma-Aldrich
Ritalinic acid, 99% (CP)
Supelco
Ritalinic acid hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®