- Immunological identification of the alternative oxidase of Acanthamoeba castellanii mitochondria.
Immunological identification of the alternative oxidase of Acanthamoeba castellanii mitochondria.
FEBS letters (1997-07-07)
W Jarmuszkiewicz, A M Wagner, M J Wagner, L Hryniewiecka
PMID9247153
ABSTRACT
Mitochondria of the protozoa Acanthamoeba castellanii possess a cyanide-insensitive oxidase cross-reacting with monoclonal antibodies raised against the plant alternative oxidase. Immunoblotting revealed three monomeric forms (38, 35, and 32 kDa) and very low amounts of a single 65 kDa dimeric form. Cross-linking studies suggest that while in plants the alternative oxidase occurs as a dimer, in amoeba it functions as a monomer. Immunologically detectable protein levels change with the age of amoeba cell culture. Increased amounts of the 35 kDa protein are accompanied by an increase in the activity of cyanide-resistant respiration.
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester, ≥98%, powder