Merck
  • Relative developmental toxicities of inhaled aliphatic mononitriles in rats.
Relative developmental toxicities of inhaled aliphatic mononitriles in rats.
Fundamental and applied toxicology : official journal of the Society of Toxicology (1993-04-01)
A M Saillenfait, P Bonnet, J P Guenier, J de Ceaurriz
ABSTRACT
The developmental toxicities of eight aliphatic mononitriles were studied in Sprague-Dawley rats after inhalation exposure for 6 hr/day, during Days 6 to 20 of gestation. The range of exposure concentrations for acetonitrile was 900 to 1800 ppm; for propionitrile and n-butyronitrile, 50 to 200 ppm; for isobutyronitrile, 50 to 300 ppm; for acrylonitrile and methacrylonitrile, 12 to 100 ppm; for allylnitrile 12 to 50 ppm; and for 2-chloroacrylonitrile, 1 to 12 ppm. Embryolethality was observed after exposure to 1800 ppm acetonitrile, 200 ppm propionitrile, 300 ppm isobutyronitrile; fetotoxicity was observed after exposure to 200 ppm propionitrile, n-butyronitrile, or isobutyronitrile, or to 25 ppm acrylonitrile in the presence of overt signs of maternal toxicity. In the absence of significant maternal toxicity, allylnitrile caused embryolethality, fetotoxicity, and clear teratogenicity at 50 ppm, and n-butyronitrile and methacrylonitrile caused fetotoxicity at 200 ppm and 100 ppm, respectively. While maternal toxicity was observed for 2-chloroacrylonitrile, it did not cause significant embryonal or fetal toxicity up to 12 ppm.
Sigma-Aldrich
Crotononitrile, mixture of cis and trans, 99%
Sigma-Aldrich
Butyronitrile, purum, ≥99.0% (GC)
Sigma-Aldrich
Butyronitrile, ≥99%