Merck
  • Functional expression of Saccharomyces cerevisiae CYP51A1 encoding lanosterol-14-demethylase in tobacco results in bypass of endogenous sterol biosynthetic pathway and resistance to an obtusifoliol-14-demethylase herbicide inhibitor.
Functional expression of Saccharomyces cerevisiae CYP51A1 encoding lanosterol-14-demethylase in tobacco results in bypass of endogenous sterol biosynthetic pathway and resistance to an obtusifoliol-14-demethylase herbicide inhibitor.
The Plant journal : for cell and molecular biology (1995-05-01)
B Grausem, N Chaubet, C Gigot, J C Loper, P Benveniste
ABSTRACT
Nicotiana tabacum protoplasts have been transformed by Agrobacterium tumefaciens containing a T-DNA in which the gene CYP51A1 encoding lanosterol-14-demethylase (LAN14DM) from Saccharomyces cerevisiae is under the control of a cauliflower mosaic virus (CaMV) 35S promoter. Two transformants strongly expressed the LAN14DM as shown by Northern and Western experiments. These transgenic calli were killed by LAB 170250F (LAB) (a phytotoxic fungicide inhibiting both plant obtusifoliol-14-demethylase (OBT14DM) and LAN14DM) but were resistant to gamma-ketotriazole (gamma-kt), a herbicide which has been shown to inhibit OBT14DM but not LAN14DM at a concentration that was lethal to control calli. However, these transgenic calli were killed by mixtures of gamma-kt plus fungicide inhibitors of LAN14DM such as ketoconazole, itraconazole or flusilazole which alone were not effective. Further analysis of the transgenic calli grown in the presence of gamma-kt showed that their delta 5-sterol content was close to that of untreated control calli obtained from protoplasts transformed with control plasmid; this is in agreement with evidence that the LAN14DM expressed from the transgene could bypass the blocked OBT14DM by using the plant substate obtusifoliol. In contrast, control calli when treated with gamma-kt, displayed a sterol content strongly enriched in 14 alpha-methyl sterols and depressed in physiological delta 5-sterols. When the transgenic calli were cultured in mixtures of gamma-kt and LAN14DM inhibitors sterol compositions enriched in 14 alpha-methyl sterols were obtained, reflecting a strong inhibition of both 'endogenous' OBT14DM and 'exogenous' LAN14DM. Taken together these results show that in tobacco calli transformed with CYP51A1, resistance to a triazole herbicide arises from expression of a functional LAN14DM enzyme; its activity in transgenic tissues creates a bypass of the sterol biosynthetic pathway at the 14-demethylase level when this latter is blocked by an OBT14DM herbicide inhibitor.
Product Description
Supelco
Flusilazole, PESTANAL®, analytical standard