Merck
  • Co-digestion of glycerine and sewage sludge to optimise green electricity production.
Co-digestion of glycerine and sewage sludge to optimise green electricity production.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-06-22)
L Maes, M Weemaes, N Hellinck, G De Gueldre, B Van De Steene
ABSTRACT
Sewage sludge and crude glycerine were co-digested in the mesophilic digester of Hoogstraten wastewater treatment plant. Additions of up to 1 kg of crude glycerine/(m³ reactor).(day) were done without significant operational problems. At higher dosage, severe digester foaming was observed. Methane production during co-digestion was up to 20% higher than what would be expected based on the digester input. Compared to the period before glycerine dosage, every tonne of added crude glycerine resulted in a surplus methane production of 489 Nm³. The theoretical methane production from the used crude glycerine was 341 Nm³ per tonne. The difference is explained by a higher sewage sludge degradability during co-digestion with glycerine. Glycerine dosage can remedy the lowered specific biogas yield of sewage sludge in Flanders and consequently enhance green electricity production.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%