Merck
  • Multicenter phase II trial of topotecan, cisplatin and bevacizumab for recurrent or persistent cervical cancer.
Multicenter phase II trial of topotecan, cisplatin and bevacizumab for recurrent or persistent cervical cancer.
Gynecologic oncology (2013-04-18)
Israel Zighelboim, Jason D Wright, Feng Gao, Ashley S Case, L Stewart Massad, David G Mutch, Matthew A Powell, Premal H Thaker, Eric L Eisenhauer, David E Cohn, Fidel A Valea, Angeles Alvarez Secord, Lynne T Lippmann, Farrokh Dehdashti, Janet S Rader
ABSTRACT
We evaluated the activity and safety of the combination of topotecan, cisplatin and bevacizumab in patients with recurrent or persistent carcinoma of the cervix. Eligible patients had persistent or recurrent cervical cancer not amenable to curative intent treatment. No prior chemotherapy for recurrence was allowed. Treatment consisted of cisplatin 50 mg/m(2) day 1, topotecan 0.75 mg/m(2) days 1, 2 and 3 and bevacizumab 15 mg/kgday 1 every 21 days until disease progression or limiting toxicity. The primary endpoint was progression free survival at 6 months. We explored PET/CT as a potential early indicator of response to therapy. Twenty-seven eligible patients received a median of 3 treatment cycles (range, 1-19). Median follow-up was 10 months (range, 1.7-33.4). The 6-month PFS was 59% (80% CI: 46-70%). In 26 evaluable patients, we observed 1 CR (4%; 80% CI: 0.4-14%) and 8 PR (31%; 80% CI: 19-45%) lasting a median of 4.4 months. Ten patients had SD (39%; 80% CI: 25-53%) with median duration of 2.2 months. Median PFS was 7.1 months (80% CI: 4.7-10.1) and median OS was 13.2 months (80% CI: 8.0-15.4). All patients were evaluated for toxicity. Grade 3-4 hematologic toxicity was common (thrombocytopenia 82% leukopenia 74%, anemia 63%, neutropenia 56%). Most patients (78%) required unanticipated hospital admissions for supportive care and/or management of toxicities. The addition of bevacizumab to topotecan and cisplatin results in an active but highly toxic regimen. Future efforts should focus on identification of predictive biomarkers of prolonged response and regimen modifications to minimize toxicity.
Sigma-Aldrich
Topotecan hydrochloride hydrate, ≥98% (HPLC and enzymatic)