Merck
  • Oxycodone + naloxone: choose morphine and optimal laxative therapy instead.
Oxycodone + naloxone: choose morphine and optimal laxative therapy instead.
Prescrire international (2013-02-02)
ABSTRACT
At high doses of oxycodone, the fixed-dose combination of oxycodone plus naloxone complicates therapeutic management and will likely have lower analgesic efficacy.
Product Description
Supelco
Oxycodone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Oxycodone hydrochloride, analytical standard