Merck
  • [Determination of patulin in juice by ultra performance liquid chromatography-tandem mass spectrometry].
[Determination of patulin in juice by ultra performance liquid chromatography-tandem mass spectrometry].
Se pu = Chinese journal of chromatography (2013-01-05)
Hua Niu, Lei Feng, Zhirui Niu, Na Zhu, Hongkun Zhu, Yaqin Wang
ABSTRACT
A method was developed for the simultaneous determination of patulin in juice by solid phase extraction (SPE)-ultra performance liquid chromatography coupled with electrospray ionization tandem mass spectrometry (UPLC-ESI-MS/MS). The cloudy juice was hydrolyzed by enzyme and extracted by ethyl acetate. The extract of cloudy juice was then enriched and purified by an HLB SPE cartridge. If the juice was clear it was directly treated by the cartridge as above. The separation was performed on a C18 column with the gradient elution of acetonitrile and water. The patulin was detected by MS with electrospray negative ionization (ESI-) in the mode of multiple reaction monitoring (MRM). The good linearity was observed in the range of 1.0 - 500 microg/L with the correlation coefficient of 0. 999 and the limit of quantitation of 5.0 microg/kg. The recoveries were in the range of 80. 6% -91. 8% at three spiked levels of 5.0, 25.0 and 100.0 microg/kg with the relative standard deviations of 1.5% -7.3%. It was proved to be a simple, reliable and accurate method which can fully meet the requirements of patulin detection in juice samples according to most domestic and international legislations.
Product Description
Supelco
Patulin solution, 100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Patulin, ≥98.0% (HPLC)