Merck
DNA analysis and sorting of viable mouse testis cells.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1981-06-01)
W M Grogan, W F Farnham, J M Sabau
ABSTRACT
The dye Hoechst 33342 and a 2-parameter cell sorter have been used to measure DNA content in viable testis cells and to sort pachytene spermatocytes and round spermatids from adult mouse testis to virtually 100% homogeneity. Early diploid spermatogenic cells were enriched to 90%, a 10-fold purification. The capability for viable sorting of most testis cell types to homogeneity in numbers suitable for many biochemical applications is demonstrated.
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥97.0% (HPLC)