- Synthesis of CdS nanoparticles in switchable surfactant reverse micelles.
Synthesis of CdS nanoparticles in switchable surfactant reverse micelles.
Chemical communications (Cambridge, England) (2012-12-14)
Jianzhong Jiang, Yane He, Leping Wan, Zhe Cui, Zhenggang Cui, Philip G Jessop
PMID23235352
ABSTRACT
A facile method for synthesis of CdS nanoparticles has been described. Monodisperse face-centered cubic CdS NPs could be synthesized in switchable surfactant reverse micelles. The micelles could be broken easily and CdS NPs without a coating of surfactants could be obtained by bubbling N(2) at 65 °C.