Merck
  • Size effects in atomic-level epitaxial redistribution process of RuO₂ over TiO₂.
Size effects in atomic-level epitaxial redistribution process of RuO₂ over TiO₂.
Scientific reports (2012-11-13)
Guolei Xiang, Xuejun Shi, Yulong Wu, Jing Zhuang, Xun Wang
ABSTRACT
Controls over the atomic dispersity and particle shape of noble metal catalysts are the major qualities determining their usability in industrial runs, but they are usually difficult to be simultaneously realized. Inspired from the Deacon catalyst in which RuO(2) can form epitaxial layers on the surfaces of Rutile TiO(2), here we have investigated the shape evolution process of RuO(2) nanoparticles on the surface of P25 TiO(2). It is found that size effects exist in this process and RuO(2) nanoparticles with sizes ~sub-2 nm can be transformed into epitaxial layers while nanoparticles with bigger sizes are not apt to change their shapes. Based on a thermodynamic model, we infer such transformation process is jointly driven by the surface tension and interfacial lattice match between the nanoparticles and substrates, which may be suggestive for the design of noble metal catalysts integrating both active crystal planes and high atomic exposure ratios.
Sigma-Aldrich
Ruthenium(IV) oxide, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium(IV) oxide hydrate, powder
Sigma-Aldrich
Ruthenium(IV) oxide hydrate, 99.9% trace metals basis