Merck
  • Thermostability, pH stability and dye degrading activity of a bacterial laccase are enhanced in the presence of Cu2O nanoparticles.
Thermostability, pH stability and dye degrading activity of a bacterial laccase are enhanced in the presence of Cu2O nanoparticles.
Bioresource technology (2012-11-08)
Arka Mukhopadhyay, Anjan Kumar Dasgupta, Krishanu Chakrabarti
ABSTRACT
The present study relates to a nanotechnology enabled method in which purified laccase from Escherichia coli AKL2 was supplemented with 100 μM copper oxide nanoparticles (Cu(2)O) (NP-laccase). The activity, half life and stability of NP-laccase were enhanced by 4, 42 and 36-fold respectively at high temperature (80 °C) and also over a wide range of pH (4-12) than laccase (in the presence of 0.18 mM CuSO(4)). Thermodynamic analysis of the nanoparticle-induced enzyme stability revealed an enhanced entropy-enthalpy compensation at 80 °C, which reflected the maintenance of its native structure. This was further supported by CD studies. The enhanced activity and thermostability of NP-laccase can be utilized for efficient decolorisation of dyes (both phenolic and azo).
Sigma-Aldrich
Copper(I) oxide, ≥99.99% trace metals basis, anhydrous
Sigma-Aldrich
Copper(I) oxide, powder, ≤7 μm, 97%