Merck
  • Divergent synthesis of novel 9-deazaxanthine derivatives via late-stage cross-coupling reactions.
Divergent synthesis of novel 9-deazaxanthine derivatives via late-stage cross-coupling reactions.
Organic & biomolecular chemistry (2012-10-11)
Francesca Bartoccini, Giovanni Piersanti, Marco Mor, Giorgio Tarzia, Patrizia Minetti, Walter Cabri
ABSTRACT
A small library of 8-substituted 9-deazaxanthines has been prepared by late-stage diversification of an 8-bromo-9-deazaxanthine. By utilizing palladium-catalyzed cross-coupling reactions a single key precursor can be transformed into a variety of 8-substituted-9-deazaxanthine compounds. Three key 8-bromo-9-deazaxanthine intermediates were efficiently prepared from commercially available 6-chlorouracil in six steps.
Sigma-Aldrich
4-Chlorouracil, ≥99%