Merck
  • Effect of salt intake on bioavailability of mizoribine in healthy Japanese males.
Effect of salt intake on bioavailability of mizoribine in healthy Japanese males.
Drug metabolism and pharmacokinetics (2012-07-21)
Kazuya Ishida, Miki Fukao, Hitomi Watanabe, Masato Taguchi, Toshio Miyawaki, Hiroyoshi Matsukura, Osamu Uemura, Zufei Zhang, Jashvant D Unadkat, Yukiya Hashimoto
ABSTRACT
Bioavailability of mizoribine in subjects with the concentrative nucleoside transporter 1 (CNT1, SLC28A1) 565-A/A allele is significantly lower than that in subjects with the SLC28A1 565-G/G allele. The aims of the present study were to investigate the cellular uptake of mizoribine in CNT1- and CNT2-expressing Madin-Darby canine kidney type II (MDCKII) cells, and to evaluate the effect of salt intake on bioavailability of mizoribine in healthy Japanese volunteers with SLC28A1 565-A/A and -G/A alleles. Eight healthy males participated in the present study, and took 150 mg mizoribine concomitantly with/without 300 mg salt. Bioavailability of mizoribine was estimated by total cumulative urinary excretion of the drug. Mizoribine was taken up Na(+)-dependently into not only CNT1-expressing but also CNT2-expressing MDCKII cells, indicating that mizoribine is a substrate for both CNT1 and CNT2. Mean bioavailability of mizoribine taken with salt (83.8%) was significantly higher than that taken without salt (73.0%). These findings suggest that the salt intake is expected to improve the bioavailability of mizoribine in patients with insufficient intestinal absorption.
Sigma-Aldrich
Mizoribine, ≥98% (TLC)