Merck
  • One pot glucose detection by [Fe(III)(biuret-amide)] immobilized on mesoporous silica nanoparticles: an efficient HRP mimic.
One pot glucose detection by [Fe(III)(biuret-amide)] immobilized on mesoporous silica nanoparticles: an efficient HRP mimic.
Chemical communications (Cambridge, England) (2012-03-27)
Bharmana Malvi, Chakadola Panda, Basab B Dhar, Sayam Sen Gupta
ABSTRACT
An [Fe(III)(biuret-amide)] complex has been immobilized onto mesoporous silica nanoparticles via Cu(I) catalyzed azide-alkyne click chemistry. This hybrid material functions as an efficient peroxidase mimic and was successfully used for the quantitative determination of hydrogen peroxide and glucose via a one-pot colorimetric assay.
Sigma-Aldrich
Biuret, 98% (Contains ≤10%(w/w) Triuret)