Merck
  • Organocatalysis as a safe practical method for the stereospecific dibromination of unsaturated compounds.
Organocatalysis as a safe practical method for the stereospecific dibromination of unsaturated compounds.
Organic letters (2012-03-27)
Gloria Hernández-Torres, Bin Tan, Carlos F Barbas
ABSTRACT
Organocatalytic stereospecific dibromination of a wide variety of functionalized alkenes was achieved using a stable, inexpensive halogen source, 1,3-dibromo 5,5-dimethylhydantoin, and a simple thiourea catalyst at room temperature. The presence of a tertiary amine enhanced the rate of the dibromination reaction, and yields were good in various solvents, including aqueous solvents. The procedure was extended to alkynes and aromatic rings and to dichlorination reactions by using the 1,3-dichloro hydantoin derivative.
Sigma-Aldrich
1,3-Dibromo-5,5-dimethylhydantoin, 98%