Merck
  • Involvement of IGF-I signaling pathway in the regulation of steroidogenesis in mouse Leydig cells treated with fenvalerate.
Involvement of IGF-I signaling pathway in the regulation of steroidogenesis in mouse Leydig cells treated with fenvalerate.
Toxicology (2012-01-03)
Jian-Hua Qu, Juan Fei, Xia Hong, Jian-Feng Chen, Ai-Hua Gu, Hong Sun, Xiao-Lin Xu, Ling Song, Shou-Lin Wang, Xin-Ru Wang
ABSTRACT
Exposure to fenvalerate has been shown to be associated with decreased steroid hormone production by mouse Leydig tumor cells (MLTC-1) in our previous study and the interference with cAMP-PKA pathway cannot explain this inhibitory effect completely. In this study, the same cell line was used to investigate the potential involvement of insulin-like growth factor I (IGF-I) signaling pathway in the downregulation of steroidogenesis by fenvalerate. Results showed that fenvalerate treatment decreased IGF-I secretion significantly which was consistent with the reduced expression of IGF-I mRNA. Then inhibitors of the two downstream pathways of IGF-I were added to the medium. The addition of LY294002 (inhibitor of phosphatidylinositol (PI)-3-kinase) did not alter the declining trend of progesterone production with increasing dosages of fenvalerate treatment while the addition of UO126 (inhibitor of extracellular signal-regulated kinases 1/2 (ERK1/2)) markedly attenuated this trend, which strongly indicated the possible involvement of pathway ERK1/2. In addition, phosphorylation of ERK1/2 was also suppressed by fenvalerate. The results suggest that the mechanism by which fenvalerate decreased steroid hormone production might involve the impairment of IGF-I signal pathway by attenuating the IGF-I production and ERK1/2 phosphorylation.
Sigma-Aldrich
Fenvalerate, ≥97%
Supelco
Esfenvalerate, PESTANAL®, analytical standard
Supelco
Fenvalerate, PESTANAL®, analytical standard