Merck
  • Nickel-catalyzed dehydrogenative [4 + 2] cycloaddition of 1,3-dienes with nitriles.
Nickel-catalyzed dehydrogenative [4 + 2] cycloaddition of 1,3-dienes with nitriles.
Journal of the American Chemical Society (2011-10-19)
Masato Ohashi, Ippei Takeda, Masashi Ikawa, Sensuke Ogoshi
ABSTRACT
Pyridines, which comprise one of the most important classes of the six-membered heterocyclic compounds, are widely distributed in nature, and the transition-metal-catalyzed [2 + 2 + 2] cycloaddition reaction of two alkynes and a nitrile is one of the most powerful methods for preparing versatile, highly substituted pyridine derivatives. However, the lack of chemo- and regioselectivity is still a crucial issue associated with fully intermolecular [2 + 2 + 2] cycloaddition. The present study developed the Ni(0)-catalyzed intermolecular dehydrogenative [4 + 2] cycloaddition reaction of 1,3-butadienes with nitriles to give a variety of pyridines regioselectively.
Sigma-Aldrich
1,3-Butadiene solution, 15 wt. % in hexane
Sigma-Aldrich
1,3-Butadiene solution, 20 wt. % in toluene