Merck
  • Total synthesis of (±)-trigonoliimine C via oxidative rearrangement of an unsymmetrical bis-tryptamine.
Total synthesis of (±)-trigonoliimine C via oxidative rearrangement of an unsymmetrical bis-tryptamine.
Journal of the American Chemical Society (2011-06-16)
Xiangbing Qi, Hongli Bao, Uttam K Tambar
ABSTRACT
We report the first total synthesis of (±)-trigonoliimine C, a member of a family of structurally complex alkaloids, in 10 steps from tryptamine and 6-methoxytryptamine. Our convergent synthetic strategy relies on a selective oxidative rearrangement of an unsymmetrical 2,2'-bis-tryptamine.
Sigma-Aldrich
Tryptamine hydrochloride, 99%
Sigma-Aldrich
Tryptamine, ≥97%