Merck
  • Removal of benzidine from polluted water by soluble and immobilized peroxidase in batch processes and continuous horizontal bed reactor.
Removal of benzidine from polluted water by soluble and immobilized peroxidase in batch processes and continuous horizontal bed reactor.
Environmental technology (2011-04-09)
Zoheb Karim, Qayyum Husain
ABSTRACT
Bitter gourd peroxidase entrapped on calcium-alginate-starch beads was employed for the treatment of water, polluted with benzidine, in a batch process as well as in a continuous reactor. The immobilized enzyme had the same pH and temperature optima as the soluble enzyme. The immobilized enzyme could effectively remove more than 70% of benzidine in a stirred batch process after three hours. After its sixth repeated use, the immobilized enzyme was able to oxidatively degrade and polymerize 58% of benzidine. The horizontal bed reactor, containing the peroxidase entrapped on calcium-alginate-starch beads, retained more than 40% benzidine removal efficiency after one month of its continuous operation. The absorption spectra of the treated benzidine exhibited a marked difference in the absorption at various wavelengths, compared with untreated compound. The horizontal bed reactor containing immobilized peroxidase would be significantly successful for the large-scale treatment of water polluted with aromatic compounds.
Sigma-Aldrich
Benzidine, ≥98.0% (N)
Sigma-Aldrich
Benzidine dihydrochloride, ≥99% (titration)
Supelco
Benzidine, analytical standard