Merck
  • A highly convergent cascade cyclization to cis-hydrindanes with all-carbon quaternary centers and its application in the synthesis of the aglycon of dendronobiloside A.
A highly convergent cascade cyclization to cis-hydrindanes with all-carbon quaternary centers and its application in the synthesis of the aglycon of dendronobiloside A.
Organic letters (2011-01-12)
Yejian Han, Lizhi Zhu, Yuan Gao, Chi-Sing Lee
ABSTRACT
An efficient and versatile ZnBr(2)-catalyzed Diels-Alder/carbocyclization cascade reaction has been developed for construction of highly functionalized cis-hydrindanes (70-96% yields with high diastereoselectivity), and it has been successfully utilized in the synthesis of the aglycon of dendronobiloside A.
Sigma-Aldrich
Zinc bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, puriss., anhydrous, ≥98%