Merck
  • D609 inhibits progression of preexisting atheroma and promotes lesion stability in apolipoprotein e-/- mice: a role of phosphatidylcholine-specific phospholipase in atherosclerosis.
D609 inhibits progression of preexisting atheroma and promotes lesion stability in apolipoprotein e-/- mice: a role of phosphatidylcholine-specific phospholipase in atherosclerosis.
Arteriosclerosis, thrombosis, and vascular biology (2010-02-09)
Lu Zhang, Jing Zhao, Le Su, Bin Huang, Li Wang, Hua Su, Yun Zhang, Shangli Zhang, Junying Miao
ABSTRACT
Atherosclerosis is considered to be a chronic inflammatory disease. Previous research has demonstrated that phosphatidylcholine-specific phospholipase C (PC-PLC) plays critical roles in various inflammatory responses. However, the association between PC-PLC and atherosclerosis is undetermined. Therefore, we sought to investigate whether PC-PLC was implicated in atherosclerosis. Immunofluorescence analysis revealed an upregulation of PC-PLC in the aortic endothelium from apolipoprotein E-deficient (apoE(-/-)) mice. PC-PLC level and activity were also increased in human umbilical vein endothelial cells in response to oxidized low-density lipoprotein treatment. Pharmacological blockade of PC-PLC by D609 inhibited the progression of preexisting atherosclerotic lesions in apoE(-/-) mice and changed the lesion composition into a more stable phenotype. Using a combination of pharmacological inhibition, polyclonal antibodies, confocal laser scanning microscopy and Western blotting, we demonstrated that PC-PLC was required for endothelial expression of lectin-like oxidized low-density lipoprotein receptor-1. In addition, D609 treatment significantly decreased the aortic endothelial expression of the vascular cell adhesion molecule-1 and the intercellular adhesion molecule-1. Furthermore, inhibition of PC-PLC in human umbilical vein endothelial cells reduced the oxidized low-density lipoprotein induced expression of vascular cell adhesion molecule-1, intercellular adhesion molecule-1, and monocyte chemotactic protein-1. Our data suggest that PC-PLC contributes to the progression of atherosclerosis.
Sigma-Aldrich
O-Tricyclo[5.2.1.02,6]dec-9-yl dithiocarbonate potassium salt, ≥95%, solid