Merck
  • Production of 4-ethylphenol from 4-hydroxycinnamic acid by Lactobacillus sp. isolated from a swine waste lagoon.
Production of 4-ethylphenol from 4-hydroxycinnamic acid by Lactobacillus sp. isolated from a swine waste lagoon.
Journal of applied microbiology (2009-12-24)
D Kridelbaugh, S Hughes, T Allen, K C Doerner
ABSTRACT
To determine the source material and the responsible micro-organisms of 4-ethylphenol production in swine lagoon sediment slurries. Swine lagoon sediment was blended and incubated with tryptone-yeast extract broth containing 10 mmol l(-1) each of 4-hydroxycinnamic as well as other phenolics. 4-Ethylphenol was only produced from 4-hydroxycinnamic acid. Denaturing gradient gel electrophoresis analysis indicated that the microbial community was substantially altered from inclusion of 4-hydroxycinnamic acid. Serial dilutions and selective plating were employed to isolate a culture capable of 4-hydroxycinnamic acid conversion to 4-ethylphenol. Morphological and ribosomal gene analysis indicated the isolate to be a Lactobacillus sp., Lactobacillus sp. pep8 converted 4-vinylphenol, but not 4-hydroxy-3-phenylpropionate, to 4-ethylphenol and did not convert 4-hydroxy-3-methoxycinnamic acid to 4-ethyl-2-methoxyphenol. A p-coumaric acid decarboxylase (pdc) gene identical to Lact. plantarum pdc genes was cloned from Lactobacillus sp. pep8. Swine lagoon sediments produce 4-ethylphenol from 4-hydroxycinnamic acid due, in part, to the activity of Lactobacillus spp. 4-Ethylphenol, a malodourant of swine and beef wastes, is generated from plant materials by indigenous lactobacilli suggesting that altering the amount dietary of plant material may influence levels of 4-ethylphenol in the wastes.
Sigma-Aldrich
Hydrocinnamic acid, 99%
Sigma-Aldrich
4-Ethylphenol, ≥98%, FG
Sigma-Aldrich
4-Ethylphenol, 99%
Sigma-Aldrich
4-Ethylphenol, ≥97.0% (GC)
Sigma-Aldrich
3-Phenylpropionic acid, 99%, FG